Learn More
Thermo Scientific™ Settling Plate Tryptic Soy Agar
Isolate and cultivate wide variety of organisms like bacteria, yeast and fungi using Settling Plate Tryptic Soy Agar.
Brand:  Thermo Scientific™ R111890
 View more versions of this product
Product Code. 17455539
  • 943.00 SEK / Pack of 10
Estimated Shipment: 03-02-2025
to see stock.
Explore more special offers
Description
Isolate and cultivate wide variety of organisms such as bacteria, yeast and fungi using Thermo Scientific™ Remel™ Settling Plate Tryptic Soy Agar. This medium is listed in Standard Methods for the Examination of Water and Wastewater and the Compendium of Methods for the Microbiological Examination of Foods1,2. This medium is recommended for use in Official Methods of Analysis of AOAC International, the International Dairy Federation (IDF), and the United States Department of Agriculture (USDA)3,4. Tryptic Soy Agar slants may be used to cultivate, store, and ship bacterial cultures.
  • Settling plates (single bag and double bag) are manufactured in a clean room environment
  • This provides the highest level of sterility assurance with no compromise of growth characteristics
  • Each lot is tested to ensure consistent, reproducible results
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Settling Plate Tryptic Soy Agar
85mm Plate, Double
Agar
10/Pk.
Not intended for IVD use.