Learn More
Lonza™ Renal Epithelial Growth Medium (REGM™) Bulletkit™
Formulated to maintain and expand most renal epithelial cells. Lonza™ Renal Epithelial Growth Medium (REGM™) Bulletkit™ contains 0.5% serum.
Brand: Lonza™ CC-3190
This item is not returnable.
View return policy
Description
- Formulated to maintain and expand most renal epithelial cells
- 1 x Bottle Basal Medium 500mL
- 1 x Green Cap Vial with hEGF 0.50mL
- 1 x Lilac Cap Vial with Insulin 0.50mL
- 1 x Natural Cap Vial with Hydrocortisone 0.50mL
- 1 x Red Cap Vial with GA-1000 0.50mL
- 1 x Bottle with FBS 2.50mL
- 1 x Natural Cap Vial with Transferrin 0.50mL
- 1 x Amber Vial with Triiodothyronine 0.50mL
- 1 x Amber Vial with Epinephrine 0.50mL
Order Info
dimProductType
Specifications
500mL bottle of REBM, (CC-3191-Renal epithelial basal medium (no growth factors) (500mL) and REGM singlequots (CC- 4127-Supplements and growth factors (hydrocortisone, hEGF, FBS, epinephrine, insulin, triiodothyronine, transferrin and gentamicin/amphotericinB). | |
Growth Medium | |
1 Kit |