Learn More
Corning™ Step-R™ Repeating Pipettor Syringe Tips, Sterile
Corning™ Step-R™ Repeating Pipettor Syringe Tips, Sterile provide precise, positive-displacement dispensing of viscous or volatile liquids.
Brand: Corning™ 6635
403.07 SEK valid until 2025-03-30
Use promo code "24134" to get your promotional price.
This item is not returnable.
View return policy
Description
- Ideal for viscous, dense and volatile liquides
- Minimum increments printed on the syringe itself
- Accurately deliver selected dose
- Made from high quality polypropylene and polyethylene
- Pyrogen-, ATP-, RNase-, and DNase-free
Specifications
Autoclavable | |
Polypropylene | |
Dispensing of viscous and volatile liquids | |
50 mL |
ISO8655 ISO11137 | |
Sterile | |
25/Pk. |