missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labconco™ Replacement Bag for Bag-In/Bag-Out HEPA/ULPA Filter
The Replacement Bag for Bag-In/Bag-Out HEPA/ULPA Filter Kit includes one clear bag and black vinyl trim.
Brand: Labconco™ 3776024
Additional Details : Weight : 3.20000kg
Description
- Two kits are required for 6 ft. wide enclosures.
Specifications
96 in. | |
Filter Accessory | |
185.4 cm |
243.8 cm | |
73 in. | |
Particulate Filtration |