Learn More
Fisherbrand™ Replacement Deuterium Lamp For Shimadzu UV and Visible HPLC Detector
Fisherbrand™ Replacement Deuterium Lamp For Shimadzu UV and Visible HPLC Detector has 2000 hours or 12 months shelf life.
Brand: Fisherbrand™ 11941-26
9929.18 SEK valid until 2025-07-13
Use promo code "24135" to get your promotional price.
Description
- Long life
- Drop-in OEM replacement
- Traceable by serial number
Order Info
Cat. No. 14-500-725 is comparable to Shimadzu part 228-34016-02.
Compliance
WEEE and ROHS compliant
Specifications
Deuterium | |
Shimadzu SPD-10A, AVP, AV, AVVP, M1 UV and Visible HPLC Detector | |
Replacement Detector Lamp |
Special Offers
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85%