Learn More
BTX™ Replacement Pins for Plate Handler
Replacement Pins designed to work with BTX™ Plate Handlers.
1320.00 SEK - 4910.00 SEK
Specifications
For Use With (Equipment) | HT 100, HT 200 |
---|---|
Material | Gold-plated |
Product Line | BTX Electroporation |
Product Type | Electroporation Spare Pin |
Electrode Shape | Pin |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15447320
|
BTX™
450468 |
25 Pk. |
1320.00 SEK
Pack of 25 |
Estimated Shipment: 22-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15457320
|
BTX™
450469 |
100 Pk. |
4910.00 SEK
Pack of 100 |
Estimated Shipment: 01-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Replacement Pins for BTX™ Plate Handlers
Specifications
HT 100, HT 200 | |
BTX Electroporation | |
Pin | |
Banana Cables | |
Yes |
Gold-plated | |
Electroporation Spare Pin | |
Non-sterile | |
Exponential Decay Wave, Square Wave |