Learn More
Moldex™ Respirator FFP 3405 P3SL Valved
The washable full face seal and unique pleated filter technology gives you the option of using these masks for more than one shift
Brand:  Moldex™ 340501
Product Code. 12322299
  • 911.00 SEK / Pack of 5
Currently Unavailable: We have no information when this product will be available
View alternative products
Explore more special offers
This item is not returnable. View return policy
Description
Description
Moldex AIR Plus 3405.Reusable FFP3 R D valved dust mask. The washable full face seal and unique pleated filter technology gives you the option of using these masks for more than one shift.
Specifications
Specifications
Respirator FFP 3405 P3SL Valved
5/Pk.