Learn More
Retsch™ Retaining Frame
Retsch™ Retaining Frame is suitable for use with Rotor Beater Mill SR 300.
Brand: Retsch™ 22.642.0004
23393.75 SEK valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Retaining frame 360°
- For use with Rotor Beater Mill SR 300
- Stainless Steel
Specifications
Retaining Frame | |
Rotor Beater Mill SR 200 | |
Stainless Steel |