Learn More
Molecular Probes™ Rhodamine Green™ Carboxylic Acid, Succinimidyl Ester, Hydrochloride (5(6)-CR 110, SE), mixed isomers
Rhodamine Green™ Carboxylic Acid, Succinimidyl Ester, Hydrochloride (5(6)-CR 110, SE), mixed isomers
Brand: Molecular Probes™ R6107
This item is not returnable.
View return policy
Description
This mixed isomer preparation of the amine-reactive Rhodamine Green carboxylic acid, succinimidyl ester yields bright and photostable conjugates. Rhodamine Green dye—labeled probes have been frequently used for Fluorescence Correlation Spectroscopy (FCS).Specifications
5 mg | |
Store in freezer (-5 to -30°C) and protect from light. | |
Room Temperature | |
Rhodamine Green™ | |
Rhodamine Green™ |
Carboxylic Acid Succinimidyl Ester | |
Amine | |
Active Ester, Succinimidyl Ester | |
Classic Dyes |
For Research Use Only. Not for use in diagnostic procedures.