missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RhoGDI Rabbit anti-Human, Mouse, Rat, Clone: 0D6X5, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Bio-Techne NBP3-15401-100UL
This item is not returnable.
View return policy
Description
RhoGDI Monoclonal antibody specifically detects RhoGDI in Human, Mouse, Rat samples. It is validated for Western Blot, KnockDownSpecifications
RhoGDI | |
Monoclonal | |
Unconjugated | |
GDIA1, MGC117248, Rho GDI 1, Rho GDP dissociation inhibitor (GDI) alpha, rho GDP-dissociation inhibitor 1, RHOGDI, Rho-GDI alpha, RHOGDI-1 | |
A synthetic peptide corresponding to a sequence within amino acids 70-150 of human RhoGDI (P52565). | |
100 μg | |
Signal Transduction | |
396 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, KnockDown | |
0D6X5 | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction