Learn More
Epredia™ Richard-Allan Scientific™ Methyl Green-Pyronin Y
Effectively identify plasma cells and RNA in tissue sections and cytological preparations with Epredia™ Richard-Allan Scientific™ Methyl Green-Pyronin Y Stain
Brand: Epredia™ 87011
This item is not returnable.
View return policy
Description
- DNA is stained green
- RNA is stained red
- Concentrated MGP is diluted prior to use
Specifications
Methyl Green-Pyronin Y | |
125mL |