Learn More
Epredia™ Richard-Allan Scientific™ Methyl Green-Pyronin Y
Effectively identify plasma cells and RNA in tissue sections and cytological preparations with Epredia™ Richard-Allan Scientific™ Methyl Green-Pyronin Y Stain
Brand:  Epredia™ 87011
Product Code. 15731209
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • DNA is stained green
  • RNA is stained red
  • Concentrated MGP is diluted prior to use
Specifications
Specifications
Methyl Green-Pyronin Y
125mL