Learn More
STEMCELL Technologies RoboSep™ Human B Cell Enrichment Kit II without CD43 Depletion
EasySep™ kit with RoboSep™ buffer and RoboSep™ filter tips
Brand:  STEMCELL Technologies 17963RF
Encompass
Product Code. 17167831
  • 16096.50 SEK / 1 set
Estimated Shipment: 02-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • The EasySep™ Human B Cell Enrichment Kit II without CD43 Depletion is designed to isolate B cells by negative selection from fresh or previously frozen PBMCs of individuals with B cell leukemia or lymphoma, or other disease states in which B cells may express CD43
  • The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles
  • The magnetically labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube
  • This kit replaces the EasySep™ Human B Cell Enrichment Kit without CD43 (Catalog #19154) for even faster cell isolations
Specifications
Specifications
B Cells
Cell Isolation
1 x 109 cells
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.