missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Rostafuroxin
Na+/K+ ATPase modulator; antihypertensive agent
2490.00 SEK - 8755.00 SEK
Specifications
Chemical Name or Material | 3β,5β,14β)-21,23-Epoxy-24-norchola-20,22-diene-3,14,17-triol |
---|---|
CAS | 156722-18-8 |
Synonym | PST 2238 |
Target | Na+/K+ ATPase Modulators |
Molecular Formula | C23H34O4 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18710268
|
Tocris Bioscience™
5315/10 |
10 mg |
2490.00 SEK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18795319
|
Tocris Bioscience™
5315/50 |
50 mg |
8755.00 SEK
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Na+/K+ ATPase modulator. Inhibits the Normalizes renal Na+/K+ ATPase pump function in mutant adducin- and endogenous ouabain-dependent forms of hypertension. Reduces blood pressure in Milan hypertensive rats. Controls mania-like behaviors in Myshkin mice having a Na+/K+ ATPase α3 subunit mutation. Antagonizes the effects of Ouabain (Cat.No. 1076) on myogenic tone and Ca2+ signaling in rat hippocampal neurons.Specifications
3β,5β,14β)-21,23-Epoxy-24-norchola-20,22-diene-3,14,17-triol | |
PST 2238 | |
C23H34O4 | |
>98% |
156722-18-8 | |
Na+/K+ ATPase Modulators | |
374.51 |
Safety and Handling
Recommended Storage : Store at 4°C