Learn More
Tocris Bioscience™ Rostafuroxin
Na+/K+ ATPase modulator; antihypertensive agent
Brand: Tocris Bioscience™ 5315/10
This item is not returnable.
View return policy
Description
Rostafuroxin is a Na+/K+ ATPase modulator. Inhibits the Normalizes renal Na+/K+ ATPase pump function in mutant adducin- and endogenous ouabain-dependent forms of hypertension. Reduces blood pressure in Milan hypertensive rats. Controls mania-like behaviors in Myshkin mice having a Na+/K+ ATPase α3 subunit mutation. Antagonizes the effects of Ouabain (Cat.No. 1076) on myogenic tone and Ca2+ signaling in rat hippocampal neurons.Specifications
3β,5β,14β)-21,23-Epoxy-24-norchola-20,22-diene-3,14,17-triol | |
156722-18-8 | |
PST 2238 | |
10 mg |