Learn More
Hettich™ Rotor Adapter for Carrier no. 1425
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
Brand:  Hettich™ 1434
 View more versions of this product
Product Code. 10272293
  • 1540.00 SEK / Each
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Rotor adapter for Hettich Carriers
  • For use with Universal 320/320 R Centrifuge
  • Universal Centrifuges can be used for both standard and special applications, they are cost-effective and save valuable space in the laboratory
Specifications
Specifications
Adapter
12 x 6 to 7 mL Tubes
100 mm
Round Bottom
12 mm
Carrier no 1425 and Swing-out Rotor 1494 for Universal 320/320 R Centrifuge