Learn More
Masterflex™ L/S™ Easy-Load™ Replacement Rotor and Bearing Support Assembly
Get the most our of your Masterflex™ I/P™ Standard Pump Head with these replacement parts
Brand: Masterflex™ 07516-08
This item is not returnable.
View return policy
Description
- Choose replacement parts suitable for your I/P™ Standard pump head size
- Suitable for Easy-Load™ Pump Heads 07516-00 and 07518-00
Specifications
Masterflex L/S Easy-Load Pump Head no. 07516-00 | |
Rotor Bearings |