Learn More
Eppendorf™ Rotor FA-30x2
Anodized aluminum rotor with an aerosol-tight QuickLock™ lid.
Brand:  Eppendorf™ 5895155006
12800.00 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15883202
  • 12851.20 SEK / Each
Explore more special offers
Description
Description
  • QuickLock™ lid
  • Aerosol-tight for safe centrifugation of hazardous samples
  • Anodized aluminum construction for high chemical resistance
  • Rotor and lid are autoclavable (121°C, 20 min.)
  • Suitable for use with 5910 R Centrifuge
TRUSTED_SUSTAINABILITY
Specifications
Specifications
30 x 1.5/2.0 mL
5910R Centrifuge
Aerosol-tight
13,700 rpm
30
Certificates
View accessory products
Product Title
Eppendorf™ Rotor FA-30x2 > 30 x 1.5/2.0 mL tubes