Learn More
Lonza™ RPMI 1640 Cell Culture Media
Brand: Lonza™ BE12-115F/U1
This item is not returnable.
View return policy
Description
Specifications
Cell Culture Media | |
2 g/L Sodium Bicarbonate, 300mg/L L-Glutamine, 5.957mg/L HEPES Buffer, 2mg/L D-Glucose anhydrous, 5mg/L Sodium Chloride, 400mg/L Potassium Chloride , 100mg/L Ca(NO3)2.4H2O , 48.83mg/L MgSO4 anhydrous , 1mg/L Glutathione reduced, 241.86mg/L L-Arginine. HCl |