missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPS10 Mouse anti-Human, Unconjugated, Clone: 1G1E1, Proteintech
Mouse Monoclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | RPS10 |
---|---|
Clone | 1G1E1 |
Concentration | 1000 μg/mL |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16840336
|
Proteintech
67609-1-IG-20UL |
20 μL |
1656.73 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16830336
|
Proteintech
67609-1-IG-150UL |
150 μL |
4721.67 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The RPS10 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RPS10. This antibody recognizes human, mouse, rat antigen. The RPS10 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
RPS10 | |
1000 μg/mL | |
Monoclonal | |
Mouse | |
6204 | |
Primary |
1G1E1 | |
Western Blot, ELISA | |
Unconjugated | |
Human, Rat, Mouse | |
IgG2b |