Learn More
RPS16 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15603-1-AP-150UL
This item is not returnable.
View return policy
Description
15603-1-AP is a rabbit polyclonal antibody raised against full-length RPS16 of human origin.
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S9P family of ribosomal proteins. It is located in the cytoplasm. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.Specifications
RPS16 | |
Polyclonal | |
Unconjugated | |
RPS16 | |
40S ribosomal protein S16, ribosomal protein S16, RPS16 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
140655, 20055, 6217 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P14131, P62249, P62250 | |
RPS16 | |
RPS16 Fusion Protein Ag7986 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |