missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RS 100329 hydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Subtype-selective α1A-adrenoceptor antagonist (pKi = 9.6 for human cloned α1A receptors). Displays 126- and 50-fold selectivity over human α1B and α1D receptors respectively. Active in vivo.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 462.89 |
Percent Purity | >99% |
Chemical Name or Material | RS 100329 hydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'