Learn More
Abeba™ Rubber 9310 Shoes
Acid and base resistant. Rubber sole provides high slip resistance.
677.14 SEK
Specifications
Certifications/Compliance | CE EN ISO 20347:2012 OB A, FO SRC/li |
---|---|
Color | Black |
Material | Leather |
Sole Features | Slip-resistant |
Sole Material | Rubber |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15643238
|
Abeba™
9310/36 |
36 |
677.14 SEK
1 pair |
Estimated Shipment: 24-10-2024
Log in to see stock. |
|||||
15653238
|
Abeba™
9310/37 |
37 |
677.14 SEK
1 pair |
Estimated Shipment: 24-10-2024
Log in to see stock. |
|||||
15663238
|
Abeba™
9310/38 |
38 |
677.14 SEK
1 pair |
Estimated Shipment: 24-10-2024
Log in to see stock. |
|||||
15673238
|
Abeba™
9310/39 |
39 |
677.14 SEK
1 pair |
Estimated Shipment: 24-10-2024
Log in to see stock. |
|||||
15683238
|
Abeba™
9310/40 |
40 |
677.14 SEK
1 pair |
Estimated Shipment: 24-10-2024
Log in to see stock. |
|||||
15693238
|
Abeba™
9310/41 |
41 |
677.14 SEK
1 pair |
Estimated Shipment: 24-10-2024
Log in to see stock. |
|||||
15603248
|
Abeba™
9310/42 |
42 |
677.14 SEK
1 pair |
Estimated Shipment: 24-10-2024
Log in to see stock. |
|||||
15613248
|
Abeba™
9310/43 |
43 |
677.14 SEK
1 pair |
Estimated Shipment: 24-10-2024
Log in to see stock. |
|||||
15623248
|
Abeba™
9310/44 |
44 |
677.14 SEK
1 pair |
Estimated Shipment: 24-10-2024
Log in to see stock. |
|||||
15633248
|
Abeba™
9310/45 |
45 |
677.14 SEK
1 pair |
Estimated Shipment: 24-10-2024
Log in to see stock. |
|||||
15643248
|
Abeba™
9310/46 |
46 |
677.14 SEK
1 pair |
Estimated Shipment: 18-10-2024
Log in to see stock. |
|||||
15653248
|
Abeba™
9310/47 |
47 |
677.14 SEK
1 pair |
Estimated Shipment: 24-10-2024
Log in to see stock. |
|||||
Description
- CE EN ISO 20347:2012 OB A, FO SRC
- Smooth leather, perforated
- Black
- PU midsole
- Slip resistance special rubber outsole sole
- Sanitized treated
- Antistatic
- Heel strap foldable, adjustable
- Leather insole
Specifications
CE EN ISO 20347:2012 OB A, FO SRC/li | |
Leather | |
Rubber |
Black | |
Slip-resistant | |
9310 |