missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ryanodine, MP Biomedicals™
Click to view available options
Quantity:
1 mg
Specifications
CAS | 15662-33-6 |
Color | White |
Molecular Formula | C25H35NO9 |
MDL Number | MFCD00151119 |
Quantity | 1 mg |
Synonym | ryanodine |
InChI Key | JJSYXNQGLHBRRK-FXKAWTGQSA-N |
SMILES | CC1CCC2(C3(CC4(C5(C(C(C3(C5(C2(C1O)O4)O)O)OC(=O)C6=CC=CN6)(C(C)C)O)C)O)C)O |
Molecular Weight (g/mol) | 493.553 |
PubChem CID | 122173044 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'