missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-3-(Boc-amino)pyrrolidine, 99%, ee 99%, Thermo Scientific Chemicals

810.00 SEK - 3017.61 SEK

Chemical Identifiers

CAS 122536-76-9
Molecular Formula C9H18N2O2
Molecular Weight (g/mol) 186.255
MDL Number MFCD00143194
InChI Key DQQJBEAXSOOCPG-ZETCQYMHSA-N
Synonym s-3-boc-amino pyrrolidine, s-tert-butyl pyrrolidin-3-ylcarbamate, s---3-boc-amino pyrrolidine, s-3-n-boc-aminopyrrolidine, tert-butyl n-3s-pyrrolidin-3-yl carbamate, s-+-3-boc-amino pyrrolidine, 3s---3-tert-butoxycarbonylamino pyrrolidine, tert-butyl s-pyrrolidin-3-ylcarbamate, s-3bocap, 3s---3-boc-amino pyrrolidine
PubChem CID 1514396
IUPAC Name tert-butyl N-[(3S)-pyrrolidin-3-yl]carbamate
SMILES CC(C)(C)OC(=O)NC1CCNC1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11325525
SDS View Documents
Thermo Scientific Alfa Aesar
L19693.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11335525
SDS View Documents
Thermo Scientific Alfa Aesar
L19693.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(S)-(-)-3-(Boc-amino)pyrrolidine, 99%, ee 99%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.