Learn More
(S)-(-)-5-Hydroxymethyl-2(5H)-furanone, 98%, Thermo Scientific Chemicals
895.00 SEK - 2350.00 SEK
Chemical Identifiers
CAS | 78508-96-0 |
---|---|
Molecular Formula | C5H6O3 |
Molecular Weight (g/mol) | 114.1 |
MDL Number | MFCD00075370 |
InChI Key | AWNLUIGMHSSXHB-BYPYZUCNSA-N |
Synonym | s---5-hydroxymethyl-2 5h-furanone, s-5-hydroxymethyl furan-2 5h-one, 2s-2-hydroxymethyl-2h-furan-5-one, 2 5h-furanone, 5-hydroxymethyl-, 5s, 5s-5-hydroxymethyl-5h-furan-2-one, pubchem7063, s-5-hydroxy-2-penten-4-olide, 5-hydroxymethyl-2 5h-furanone #, 5beta-hydroxymethyl furan-2 5h-one |
PubChem CID | 10176122 |
IUPAC Name | (2S)-2-(hydroxymethyl)-2H-furan-5-one |
SMILES | C1=CC(=O)OC1CO |
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
11312717
|
Thermo Scientific Alfa Aesar
L11919.MD |
250 mg |
895.00 SEK
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11302717
|
Thermo Scientific Alfa Aesar
L11919.03 |
1 g | N/A | N/A | N/A | |||||
Description
(S)-(-)-5-Hydroxymethyl-2(5H)-furanone is used in the preparation of partially saturated heterocycles via a diastereoselective ring chain formation. It is also used in the synthesis of (1R,3S)-cis-chrysanthemic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(S)-(-)-5-Hydroxymethyl-2(5H)-furanone is used in the preparation of partially saturated heterocycles via a diastereoselective ring chain formation. It is also used in the synthesis of (1R,3S)-cis-chrysanthemic acid.
Solubility
Slightly soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Chemical Identifiers
78508-96-0 | |
114.1 | |
AWNLUIGMHSSXHB-BYPYZUCNSA-N | |
10176122 | |
C1=CC(=O)OC1CO |
C5H6O3 | |
MFCD00075370 | |
s---5-hydroxymethyl-2 5h-furanone, s-5-hydroxymethyl furan-2 5h-one, 2s-2-hydroxymethyl-2h-furan-5-one, 2 5h-furanone, 5-hydroxymethyl-, 5s, 5s-5-hydroxymethyl-5h-furan-2-one, pubchem7063, s-5-hydroxy-2-penten-4-olide, 5-hydroxymethyl-2 5h-furanone #, 5beta-hydroxymethyl furan-2 5h-one | |
(2S)-2-(hydroxymethyl)-2H-furan-5-one |
Specifications
78508-96-0 | |
C5H6O3 | |
3537455 | |
s---5-hydroxymethyl-2 5h-furanone, s-5-hydroxymethyl furan-2 5h-one, 2s-2-hydroxymethyl-2h-furan-5-one, 2 5h-furanone, 5-hydroxymethyl-, 5s, 5s-5-hydroxymethyl-5h-furan-2-one, pubchem7063, s-5-hydroxy-2-penten-4-olide, 5-hydroxymethyl-2 5h-furanone #, 5beta-hydroxymethyl furan-2 5h-one | |
AWNLUIGMHSSXHB-BYPYZUCNSA-N | |
C1=CC(=O)OC1CO | |
114.1 | |
114.1 | |
(S)-(-)-5-Hydroxymethyl-2(5H)-furanone |
34°C to 43°C | |
MFCD00075370 | |
Hygroscopic | |
Slightly soluble in water. | |
−148° (c=1.4 in Water) | |
(2S)-2-(hydroxymethyl)-2H-furan-5-one | |
10176122 | |
98% |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.