Learn More
Fisherbrand™ S-Series Heated Ultrasonic Cleaning Bath
With state of the art microprocessor controlled ultrasonic cleaning and sweep technology. Fisherbrand™ S-Series Ultrasonic Cleaners offer a comprehensive range of analogue ultrasonic water baths covering the requirements for laboratory research, sonochemistry and degassing applications.
3502.00 SEK - 54370.00 SEK
Product Code | Brand | Capacity (Metric) | Dimensions (D x W x H) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Dimensions (D x W x H) | Price | Quantity & Availability | ||||
10591203
|
Fisherbrand™
1001214 |
0.8 L | 116 x 206 x 178 mm | N/A | |||||
10551783
|
Fisherbrand™
1002060 |
1.75 L | 180 x 175 x 212 mm | N/A | |||||
10162372
|
Fisherbrand™
1001955 |
2.75 L | 179 x 300 x 214 mm | N/A | |||||
10215332
|
Fisherbrand™
1002006 |
4.25 L | 179 x 300 x 264 mm | N/A | |||||
10345822
|
Fisherbrand™
1002238 |
5.75 L | 186 x 365 x 264 mm | N/A | |||||
10395912
|
Fisherbrand™
1002140 |
6.9 L | 179 x 568 x 214 mm | N/A | |||||
10663113
|
Fisherbrand™
1002170 |
9.4 L | 179 x 568 x 264 mm | N/A | |||||
10239491
|
Fisherbrand™
1002279 |
9.5 L | 278 x 365 x 264 mm | N/A | |||||
10204762
|
Fisherbrand™
1002825 |
12.75 L | 278 x 365 x 321 mm | N/A | |||||
10102762
|
Fisherbrand™
1002877 |
18 L | 340 x 390 x 321 mm | N/A | |||||
15280147
|
Fisherbrand™
1071162 |
28 L | 340 x 568 x 321 mm | N/A | |||||
10366012
|
Fisherbrand™
1003908 |
45 L | 370 x 615 x 467 mm | N/A | |||||
10427912
|
Fisherbrand™
1003946 |
90 L | 570 x 715 x 467 mm | N/A | |||||
Description
- Long lifespan from cavitation resistant stainless steel tanks
- LED display showing set and remaining time and pre-set and actual temperature
- Newly developed cover reduces noise and accelerates the heating up process
- Easy draining of cleaning liquid though the drain duct via a dial on the side
- Operating panel arranged so that liquid cannot enter the electronics
- Degas function key
Specifications
30°C to 80°C |