missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S1P5/EDG-8 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
1680.00 SEK - 3415.00 SEK
Specifications
Antigen | S1P5/EDG-8 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18323062
|
Bio-Techne
NBP3-15965-20UL |
20 μg |
1680.00 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18369075
|
Bio-Techne
NBP3-15965-100UL |
100 μg |
3415.00 SEK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
S1P5/EDG-8 Polyclonal antibody specifically detects S1P5/EDG-8 in Human samples. It is validated for Western BlotSpecifications
S1P5/EDG-8 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
Edg-8, EDG8S1P receptor Edg-8, Endothelial differentiation G-protein-coupled receptor 8, endothelial differentiation, sphingolipid G-protein-coupled receptor, 8, S1P receptor 5, S1P5, sphingosine 1-phosphate receptor 5, sphingosine 1-phosphate receptor EDG8, Sphingosine 1-phosphate receptor Edg-8, sphingosine-1-phosphate receptor 5, SPPR-1, SPPR-2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 307-398 of human S1P5/EDG-8 (NP_110387.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
GPCR | |
PBS, 50% glycerol, pH7.3 | |
53637 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title