missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Scharlab Sabouraud Chloramphenicol Agar
Scharlab Sabouraud Chloramphenicol Agar for the isolation of fungi.
Brand: Scharlab 01-166-500
This item is not returnable.
View return policy
Description
- EP/ISO
- Solid culture medium for the isolation of fungi
- According to ISO 16212 and 18416 standards.
Specifications
ISO 16212 and 18416 | |
Sabouraud Chloramphenicol Agar | |
Casein Peptone 5.0 g/L, Meat Peptone 5.0 g/L, D(+)-Glucose 40.0 g/L, Chloramphenicol 0.5 g/L, Agar 15.0 g/L | |
20°C to 25°C | |
5.6 ±0.2 |
Solid | |
Microbiology Media | |
Isolation of Fungi | |
48 hr. to 5 Days | |
500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction