Learn More
Notrax Safety Stance Solid™ High Quality Anti-Fatigue Matting
High quality anti-fatigue matting for heavy duty industrial environments. Unique DiamondGrid™ surface pattern ensures sure footing to prevent slips and falls while channels drain incidental oil exposure.
Brand:  Notrax 649S3124OB
 View more versions of this product
Product Code. 17752248
  • 10910.00 SEK / Each
Estimated Shipment: 22-01-2025
to see stock.
Explore more special offers
Description
  • High quality anti-fatigue matting for heavy duty industrial environments
  • Unique DiamondGrid™ surface pattern ensures sure footing to prevent slips and falls while channels drain incidental oil exposure
  • Closed design ensures easy cleaning and prevents small items from falling under the mat
  • Made of 100% nitrile rubber compound designed to withstand industrial oils and harsh chemicals
  • Suited for multi-shift operations
  • Remarkable worker comfort derived from 20 mm thick resilient worker platform
  • Standard size mats for individual workstations are loose lay and require no installation
  • Highly visible nitrile rubber orange colored borders included on 3 sides for added safety to prevent tripping
  • Non-shifting
  • Free of silicone therefore safe for vehicle painting facilities
Specifications
Specifications
Black/Orange
315 cm
Industrial and Commercial Matting
10.3 ft.
Nitrile Rubber
97 cm