Learn More
SymLine™ Safety Waste Caps
These Safety Waste Caps safely connect the pipework to the waste container as an integrated exhaust filter blocks solvent vapors for health and environmental protection.
Brand: SymLine™ 106718
5359.74 SEK valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Available with different thread sizes for attachment to pipe system
- Different numbers of hose connections for disposal of liquid waste
- Available with electronic level sensors that indicate the degree of filling of the disposal container
- By using a 4-in-1 collector (306509), up to 4 Safety Waste Caps can be connected simultaneously to only one connection point on the pipe system
Specifications
Black | |
SymLine System | |
PTFE-EC | |
Safety Waste Cap |
Waste Disposal | |
Electronic Level Control with Shut-off GL25 (m), Connector for Exhaust Filter | |
S60/61 |