missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Salicin, MP Biomedicals
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
Specifications
Specifications
Quantity | 5 g |
Formula Weight | 286.3 |
Physical Form | Powder |
Chemical Name or Material | Salicin |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'