missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ Sampler Rack

These Sampler Racks are ideal for hanging up samplers when they are not in use.

Brand:  Sampling Systems™ 8820A-01

 View more versions of this product

Product Code. 15899561

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Stainless steel and plastic construction
  • Standard rack can hold up to 5 samplers
  • Samplers are attached by a simple push clip design
  • Comes complete with hole fixings
  • Suitable for samplers from 10 to 35mm in diameter and up to 4kg in weight
Specifications

Specifications

Storage Rack
Stainless Steel, Plastic
Sampler storage
900 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sampling Systems™ Sampler Rack > 900mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.