missing translation for 'onlineSavingsMsg'
Learn More

Saint Gobain Life Sciences™ Sani-Tech™ STHT™-R Braid-reinforced Silicone Tubing

This braid-reinforced, platinum-cured silicone hose is an ultra-flexible, high-purity hose that was developed for higher-pressure applications.

Brand:  Saint Gobain Life Sciences™ STHT-R-0375

 View more versions of this product

Product Code. 17953831

  • 10596.89 SEK / 7.50m

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Manufactured with Sani-Tech™ 65 custom-brand silicone resin
  • Resists temperature extremes, ozone, radiation, moisture, compression sets, weathering, and chemical attack
  • Imparts no taste or odors to fluids transported within it
  • Withstands repeated autoclaving and sterilization
  • Resists the adherence of blood products and other sanitary fluids
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Tubing
0.374 in.
Silicone
15.9 mm
7.5 m
3.2 mm
326 lbf/in.
65 (Shore A)
180 psi
260°C
Autoclavable
9.5 mm
0.626 in.
24.6 in.
0.126 in.
Pharmaceutical Silicone Tubing
Clear
50 mm bend radius
500°F
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers