Learn More
Notrax Sani-Trax™ Disinfecting Foot Bath Entrance Mat, Black
Notrax Sani-Trax™ Disinfecting Foot Bath Entrance Mat cleans and sanitizes shoes, recommended at entrances to all food processing areas. As traffic passes, rubber tips bend to immerse soles in disinfectant solution, a double action that keeps contamination from reaching food processing areas.
455.00 SEK - 643.00 SEK
Specifications
Color | Black |
---|---|
Material | Natural Rubber |
Type | Industrial and Commercial Matting |
Product Code | Brand | Length (Metric) | Width (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Width (Metric) | Price | Quantity & Availability | ||||
17782018
|
Notrax
346S1824BL |
24 cm | 18 cm |
455.00 SEK
Each |
Estimated Shipment: 09-07-2024
Log in to see stock. |
||||
17792018
|
Notrax
346S2432BL |
80 cm | 60 cm |
643.00 SEK
Each |
Estimated Shipment: 09-07-2024
Log in to see stock. |
||||
Description
- This mat cleans and sanitizes shoes, recommended at entrances to all food processing areas
- Thousands of flexible fingers provide an automatic scrubbing action that dislodges contaminants from footwear
- As traffic passes, rubber tips bend under pressure to immerse soles in disinfectant solution, a double action that keeps contamination from reaching food processing areas
- Convenient mat size fits most doorways
- Suction cups on underside prevents mat from shifting
Specifications
Black | |
Natural Rubber | |
Industrial and Commercial Matting |