Learn More
Sartorius Funnel
Reusable stainless steel funnels for microbial enumeration and particle counting.
Brand: Sartorius 6981065
4103.20 SEK valid until 2024-12-31
Use promo code "23270" to get your promotional price.
Additional Details : Weight : 0.10000kg
Description
- Specially designed for use in microbiological quality assurance, these three filter holders differ only in their capacity
- The reusable funnels feature a special locking clamp that simplifies the positioning and removal of the membrane filter, ensuring secure lock-down
- For traceability, each filter funnel has an individual serial number
Specifications
Autoclavable | |
Combisart System | |
Colony or particle counting | |
Stainless Steel |
100 mL | |
Funnel | |
Closure clamp | |
4.7/5 cm |