Learn More
Sartorius Polyethersulfone Membrane Filters
Polyethersulfone (PES) filters have a high internal porosity.
Brand: Sartorius 15458--50------N
2712.25 SEK valid until 2024-12-31
Use promo code "21931" to get your promotional price.
Additional Details : Weight : 0.10000kg
Description
Polyethersulfone (PES) filters have a high internal porosity.
They perform well at high flux with an excellent throughput of aqueous solutions over the entire pH range of 1-14. Thanks to their low non-specific protein adsorption the PES type is recommended for filtering biological and pharmaceutical solutions.
Specifications
PES | |
Recommended for filtering biological and pharmaceutical solutions. | |
150 μm |
0.1 μm | |
100/Pk. | |
50 mm |