missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SB 206553 hydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent and selective 5-HT2B/5-HT2C receptor antagonist (rat 5-HT2B pA2 = 8.89, human 5-HT2C pKi = 7.92). Displays > 80-fold selectivity over all other 5-HT receptor subtypes and a variety of other receptors (pKi < 6). Centrally active following oral administration in vivo.
Specifications
Quantity | 10 mg |
Synonym | sb 206553 hydrochloride, n-3-pyridinyl, dsstox_cid_22622, dsstox_rid_80061, dsstox_gsid_42622, sb206553 hydrochloride, 3,5-dihydro-5-methyl-n-3-pyridinylbenzo 1,2-b:4,5-b' dipyrrole-1 2h-carboxamide hydrochloride, 3,5-dihydro-5-methyl-benzo 1,2-b:4,5-b' dipyrrole-1 2h-carboxamide hydrochloride, 5-methyl-1-3-pyridylcarbamoyl-1,2,3,5-tetrahydropyrrolo 2,3-f indole hydrochloride, 5-methyl-n-pyridin-3-yl-3,5-dihydropyrrolo 2,3-f indole-1 2h-carboxamide hydrochloride |
Formula Weight | 328.8 |
Percent Purity | >99% |
Chemical Name or Material | SB 206553 hydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'