missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SB 408124, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
Description
Selective non-peptide orexin OX1 receptor antagonist (Kb values are 21.7 and 1405 nM for human OX1 and OX2 receptors respectively). Blocks orexin-A induced grooming following oral administration in vivo.
Specifications
CAS | 288150-92-5 |
Molecular Formula | C19H18F2N4O |
Quantity | 10 mg |
Synonym | n-6,8-difluoro-2-methyl-4-quinolinyl-n'-4-dimethylamino phenyl urea, 1-6,8-difluoro-2-methylquinolin-4-yl-3-4-dimethylamino phenyl urea, 1-6,8-difluoro-2-methylquinolin-4-yl-3-4-dimethylaminophenyl urea, acmc-20p1d0, d0j3pj, sb hplc , solid, 3-6,8-difluoro-2-methylquinolin-4-yl-1-4-dimethylamino phenyl urea, n-6,8-difluoro-2-methyl-4-quinolinyl-n'-4-dimethylamino phenyl-urea |
InChI Key | JTARFZSNUAGHRB-UHFFFAOYSA-N |
SMILES | CC1=NC2=C(C=C(C=C2C(=C1)NC(=O)NC3=CC=C(C=C3)N(C)C)F)F |
IUPAC Name | 1-(6,8-difluoro-2-methylquinolin-4-yl)-3-[4-(dimethylamino)phenyl]urea |
Molecular Weight (g/mol) | 356.377 |
PubChem CID | 4331799 |
Formula Weight | 356.37 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'