Learn More
Buerkle™ Brass Screw Joint for Barrel Pump
Buerkle™ Brass Screw Joint for Barrel Pump prevents electro-static charge in combination with the anti-static set.
Brand: Buerkle™ 5601-0100
621.57 SEK valid until 2025-03-31
Use promo code "24119" to get your promotional price.
This item is not returnable.
View return policy
Description
- Barrel screw joint made of nickel-plated brass for barrel pump stainless steel
- Prevents electro-static charge in combination with the Anti-static set
- For the safe transfer of flammable and easily flammable liquids
- For containers with R 2 in. steel fine thread inside
- The container connectors make decanting even easier and safer with their more secure connection to barrels
- The connectors secure the pump in the barrel, enable a vertical position change and allow the required air exchange
Specifications
Stainless Steel Barrel Pump | |
Nickel-plated Brass | |
Barrel Screw Joint |
Product Content Correction
Product Title
Buerkle™ Brass Screw Joint for Barrel Pump > Material: Nickel-plated brass
Spot an opportunity for improvement?Share a Content Correction