Learn More
BD Select APS Tryptic Soy Broth
BD Select APS Tryptic Soy Broth is used in microbiology laboratories as a culture broth to grow bacteria.
Brand:  BD 214886
Product Code. 17362301
  • 49290.00 SEK / Each
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Tryptic soy broth is used in microbiology laboratories as a culture broth to grow aerobic bacteria.
    • Used in a wide variety of procedures
    • May be used for suspension and enrichment
    • For cultivation of strains isolated on other media
  • With the addition of 6.5% sodium chloride, differentiation between salt-tolerant and salt-intolerant species is achieved.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TSB
Blue
10 kg