missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WTW™ SenTix™ ORP Electrodes
Different metal electrodes to adapt to measured substance, Platinum ORP electrodes are suitable for the most varied applications: strongly oxidizing solutions, argentometry and much more.
Brand: WTW™ 103648
2584.33 SEK valid until 2024-12-31
Use promo code "21658" to get your promotional price.
This item is not returnable.
View return policy
Description
- Platinum, gold or silver ORP electrodes - suitable for the most varied applications: strongly oxidizing solutions, argentometry and much more
- Different metal electrodes to adapt to measured substance
- Plug head electrodes for universal use
- Ag/AgCl reference system
Specifications
Glass Shaft, Ceramic Diaphragm, Platinum Sensor | |
SenTix™ ORP Electrode | |
SenTix™ ORP | |
120 mm |
0°C to 100°C | |
ORP Measurement meter | |
0°C to 100°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction