missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Graduated Separating Funnel

Increase precision control of flow with a needle control funnel. Kartell™ Graduated Separating Funnel features detachable attachments for easy cleaning.

Brand:  Kartell™ 0097800

548.26 SEK valid until 2024-11-29
Use promo code "21718" to get your promotional price.


Product Code. 10672702

  • 856.00 SEK / Each
Estimated Shipment: 07-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Crystal clear funnel
  • Permanent graduations moulded into body
  • Needle control for precise control of flow
  • No wetting
  • Top unscrews and valve can be detached for thorough cleaning
  • Autoclavable
  • Excellent chemical resistance
  • Complete with stopper
Specifications

Specifications

500 mL
Autoclavable
2mL, 5mL, 25mL
PMP
Clear
NS DIN 29/32
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers