missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SERPINA3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
1422.00 SEK - 4180.00 SEK
Specifications
Antigen | SERPINA3 |
---|---|
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17763644
|
Invitrogen
PA5122248 |
20 μL |
1422.00 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17753644
|
Invitrogen
PA5122247 |
100 μL |
4180.00 SEK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SERPINA encodes Alpha 1-ACT. It is an early-stage acute-phase plasma protein and a serpin that preferentially inactivates chymotrypsin, cathepsin G, and chymase. Alpha-1-ACT, a serine protease inhibitor, is tightly associated with amyloid plaques in Alzheimer's disease (AD) and in normal aged human and monkey brain. Regulation of the serine proteases and serine protease inhibitors plays an important role in neuromuscular differentiation. Prostate specific antigen (PSA), a chymotrypsin-like serine protease, is predominantly complexed to Alpha-1-ACT.Specifications
SERPINA3 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P01011 | |
12 | |
Lys235-Arg394 (Accession P01011), with N-terminal His and GST Tag | |
Primary | |
-20°C | |
SERPINA3 |
1 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
AACT; ACT; alpha-1-antichymotrypsin; Alpha-1-antichymotrypsin His-Pro-less; Cell growth-inhibiting gene 24/25 protein; GIG24; GIG25; Growth inhibiting protein 24; Growth inhibiting protein 25; growth-inhibiting protein 24; growth-inhibiting protein 25; MGC88254; Serine (or cysteine) proteinase inhibitor clade A member 3; serine (or cysteine) proteinase inhibitor, clade A, member 3; serpin A3; serpin family A member 3; Serpin peptidase inhibitor clade A (alpha 1 antiproteinase antitrypsin) member 3; serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 3; SERPINA3 | |
SERPINA3 | |
IgG | |
Antigen affinity chromatography | |
Antibody |