missing translation for 'onlineSavingsMsg'
Learn More

SETD5 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

1935.00 SEK - 4555.00 SEK

Specifications

Antigen SETD5
Applications Immunohistochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18346524
View Documents
Bio-Techne
NBP3-05089-20UL
20 μL
1935.00 SEK
20µL
Please to purchase this item. Need a web account? Register with us today!
18332243
View Documents
Bio-Techne
NBP3-05089-100UL
100 μL
4555.00 SEK
100µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SETD5 Polyclonal specifically detects SETD5 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

SETD5
Polyclonal
Purified
RUO
PBS with 50% glycerol, pH7.3.
55209
IgG
Affinity purified
Immunohistochemistry, Immunofluorescence
Unconjugated
Rabbit
Human
DKFZp686J18276, FLJ10707, KIAA1757, SET domain containing 5, SET domain-containing protein 5
Recombinant fusion protein containing a sequence corresponding to amino acids 520-740 of human SETD5 (NP_001073986.1).
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.