missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SETD7/9 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
1670.00 SEK - 3970.00 SEK
Specifications
Antigen | SETD7/9 |
---|---|
Dilution | Western Blot 1:1000 |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18358003
|
Bio-Techne
NBP3-18644-25UL |
25 μg |
1670.00 SEK
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18323795
|
Bio-Techne
NBP3-18644-100UL |
100 μg |
3970.00 SEK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SETD7/9 Polyclonal antibody specifically detects SETD7/9 in Human samples. It is validated for Western BlotSpecifications
SETD7/9 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Histones and Modified Histones, Signal Transduction | |
Whole antiserum | |
EC 2.1.1.43, H3-K4-HMTase SETD7, histone H3-lysine 4-specific methyltransferase, histone-lysine N-methyltransferase SETD7, KIAA1717SET domain-containing protein 7, KMT7FLJ21193, Lysine N-methyltransferase 7, SET domain containing (lysine methyltransferase) 7, SET7/9Histone H3-K4 methyltransferase SETD7, SET7Set9, SET9 | |
Polyclonal antibody raised in rabbit against human SETD7/9 (SET domain containing (lysine methyltransferase) 9), using a recombinant protein. | |
Whole Antiserum |
Western Blot 1:1000 | |
Polyclonal | |
Antisera | |
RUO | |
Human | |
Q8WTS6 | |
80854 | |
Primary | |
Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title