missing translation for 'onlineSavingsMsg'
Learn More
Learn More
sFRP-1 Rabbit anti-Human, Mouse, Rat, Clone: 5I6V1, Novus Biologicals™
Rabbit Monoclonal Antibody
1905.00 SEK - 4805.00 SEK
Specifications
Antigen | sFRP-1 |
---|---|
Clone | 5I6V1 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18323543
|
Bio-Techne
NBP3-16851-20UL |
20 μg |
1905.00 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18368546
|
Bio-Techne
NBP3-16851-100UL |
100 μg |
4805.00 SEK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
sFRP-1 Monoclonal antibody specifically detects sFRP-1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
sFRP-1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Apoptosis | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
6422 | |
IgG | |
Affinity purified |
5I6V1 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
FRP1, FRP-1FrzA, FRPSARP-2, SARP2sFRP-1, Secreted apoptosis-related protein 2, secreted frizzled-related protein 1 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human sFRP-1 (Q8N474). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title