missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trajan™ Phenyl Deactivated Fused Silica Capillary Tubing
Essential for maintaining a high performance chromatography system. Trajan Scientific and Medical™ Phenyl Deactivated Fused Silica Capillary Tubing is suitable for organic and aqueous solvents
Brand: Trajan™ 064114
This item is not returnable.
View return policy
Description
- Chemically inert and thermally stable to 380°C
- Suitable for organic and aqueous solvents
- Ideal for biotechnology applications
Specifications
Capillary Tubing | |
0.53 mm | |
Phenyl |
0.68 mm | |
Organic and Aqueous Solvents | |
5 m |