Learn More
SGE™ GC Inlet FocusLiners™
Results in lower mass discrimination
Brand:  SGE™ 092002
 View more versions of this product
Product Code. 10592393
  • 2010.00 SEK / Pack of 5
Estimated Shipment: 13-02-2025
to see stock.
Explore more special offers
Description
Results in lower mass discrimination
  • For various GC Instruments
  • All liners are deactivated using high temperature (420°C) gas phase deactivation
  • Supplied in blister packs to prevent contamination
  • Reduced active sites mean no unwanted reaction in samples
FocusLiner
  • Includes two tapered sections that secure quartz wool plug for optimum needle tip wiping and sample transfer, also prevents quartz wool from shifting toward septum or pushing down into liner
  • Provide excellent reproducibility
Specifications
Specifications
5/Pk.
Quartz Wool
0.4 cm
Agilent HP 5890, 6890, 7890, 6850, 4890, Thermo 9001, GCQ TRACE 1300, Varian 1177
7.85 cm
Split/Splitless Fast Focus Liner
0.63 cm