Learn More
Eurofins DiscoverX SGK3 Protein, Unactive
Unactive, amino acid residues 119-end, for use in Kinase Assays.

6046.30 SEK - 46147.75 SEK
Specifications
Formulation | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
---|---|
Molecular Weight (g/mol) | 46.8 kDa |
Species | Human |
Recombinant | Recombinant |
Protein Tag | N-terminal 6His |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17320494
![]() |
Eurofins DiscoverX
14-648 |
10 μg |
6046.30 SEK
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17330494
![]() |
Eurofins DiscoverX
14-648M |
250 μg |
46147.75 SEK
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SGK3 is a member of the Ser/Thr protein kinase family and encodes a phosphoprotein with a PX (phox homology) domain. The protein phosphorylates several target proteins and has a role in neutral amino acid transport and activation of potassium and chloride channels. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
GenBank NM_013257
Specifications
mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. | |
Human | |
N-terminal 6His | |
Truncated | |
SGK3, unactive |
46.8 kDa | |
Recombinant | |
Recombinant enzyme expressed in Sf21 insect cells | |
≥73% |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.