Learn More
Epredia™ Shandon™ Dual Blade Manual Bone Saw: Replacement Blades
Replacement Blades for use with Shandon Dual Blade Manual Bone Saw.
Brand: Epredia™ 1001798
463.25 SEK valid until 2024-12-31
Use promo code "21628" to get your promotional price.
This item is not returnable.
View return policy
Description
High-quality blades for use with Shandon Dual Blade Manual Bone SawSpecifications
Replacement Blades, Coarse | |
Replacement blade set (includes links) | |
17.8 cm | |
22.9 cm |
Dual Blade Manual Bone Saw (1001797) | |
7 in. | |
9 in. |